Learn More
Apexbio Technology LLC Fluticasone propionate, 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B20965.1
Encompass_Preferred
Fluticasone propionate is a high affinity, selective GR (glucocorticoid receptor) agonist which is derived from fluticasone used to treat asthma and allergic rhinitis.
Catalog No. NC3343323