Learn More
Medchemexpress LLC Fluticasone (propionate) | 80474-14-2 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0154100MG
Encompass_Preferred
Fluticasone propionate is a potent topical anti-inflammatory corticosteroid and a selective glucocorticoid receptor agonist, with an absolute affinity (KD) of 0.5 nM. It shows little or no activity at other steroid receptors and also has anti-viral activity.
  • Potent topical anti-inflammatory corticosteroid.
  • Selective glucocorticoid receptor agonist (KD of 0.5 nM).
  • Shows little or no activity at other steroid receptors.
  • Has anti-viral activity.
  • Inhibits TNFα-induced E-selectin expression (IC50 of 1 nM).
  • Targets: Glucocorticoid Receptor, Endogenous Metabolite, Enterovirus, and ADC Payload.
Catalog No. 50-004-07195