Learn More
Medchemexpress LLC Fluticasone propionate | 80474-14-2 | 99.93% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0154
Encompass_Preferred
Fluticasone propionate is a potent topical anti-inflammatory corticosteroid and a selective glucocorticoid receptor agonist with an absolute affinity (KD) of 0.5 nM. It exhibits little or no activity at other steroid receptors and also shows anti-viral activity.
  • Inhibits TNFα-induced E-selectin expression with an IC50 of 1 nM.
  • Shows EC50 of 0.7 nM for transactivation activity at the glucocorticoid receptor in HepG2 cells.
  • Exhibits ED50 of 3 μg for antiinflammatory activity in neutrophils.
  • Provides IC50 of 0.1 nM for inhibition of TNFalpha release in PBMC.
  • Demonstrates EC50 of 1.09 nM for agonist activity at GR in SW1353 cells.
Catalog No. 50-004-05807