Learn More
Medchemexpress LLC Fluticasone propionate | 80474-14-2 | MFCD00866007 | 99.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB015410MM/1ML
Encompass_Preferred
Fluticasone propionate is a potent topical anti-inflammatory corticosteroid that acts as a selective glucocorticoid receptor agonist, with an absolute affinity (KD) of 0.5 nM. It exhibits little to no activity at other steroid receptors and also possesses anti-viral activity. This product is intended for research use only.
  • Available as solid and a 10 mM solution in DMSO.
  • Shipped at room temperature within the continental US.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
  • Soluble in DMSO at 100 mg/mL (199.77 mM).
  • Insoluble in H2O.
Catalog No. 50-004-06236