Learn More
Medchemexpress LLC Flutriafol | 76674-21-0 | MFCD00144319 | 95.0% | 301.29 g·mol⁻¹ | C16H13F2N3O | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW019852
Encompass_Preferred
Flutriafol is a triazole-class fungicide provided as an analytical standard for research and analytical applications. It is used for identification and quantification in analytical assays, method development, and studies of fungicidal activity. Intended for research use only; not for human therapeutic use.
  • Triazole fungicide with broad-spectrum activity.
  • Suitable as an analytical standard for quantitative assays and method development.
  • High chemical purity for reliable analytical results.
  • Stable powder form with recommended cold storage for long-term stability.
  • Available in small package sizes suitable for laboratory workflows.
Catalog No. 50-003-03190