Learn More
Medchemexpress LLC Flutriafol | 76674-21-0 | MFCD00144319 | 98.0% | 301.29 g/mol | C16H13F2N3O | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01985210MM/1ML
Encompass_Preferred
Flutriafol is a triazole-class fungicide supplied for laboratory research and analytical applications. It exhibits broad-spectrum fungicidal activity and is provided as solid material or as ready-to-use DMSO solutions. Products are intended for research use only and are not for human use.
  • Broad-spectrum fungicidal activity against plant pathogens.
  • Triazole mode of action targeting fungal sterol biosynthesis.
  • Offered as ready-to-use 10 mM solution in DMSO and as solid material.
  • High purity suitable for analytical and research applications (~98.0% purity).
  • Multiple package sizes available to support varied testing needs.
  • Intended for research use only; not for human use.
Catalog No. 50-003-01701