Learn More
Medchemexpress LLC Flutriafol | 76674-21-0 | MFCD00144319 | 98.0% | 301.29 g·mol⁻¹ | C16H13F2N3O | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW01985250MG
Encompass_Preferred
Flutriafol is a triazole-class fungicide supplied as an analytical standard for research and analytical applications. It exhibits broad-spectrum fungicidal activity and is provided with supporting analytical data and handling instructions for laboratory use.
  • Triazole-class fungicide with broad-spectrum activity.
  • Supplied as a solid and as DMSO solutions for analytical work.
  • High reported purity (98.02%).
  • Characterized by molecular weight 301.29 g·mol⁻¹ and formula C16H13F2N3O.
  • Intended for research use only with accompanying SDS and COA documentation.
  • Storage guidance provided for powder and solution forms.
Catalog No. 50-003-00104