Learn More
TARGETMOL CHEMICALS INC FLUTRIMAZOLE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T611485MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Flutrimazole is a dual-action imidazole antifungal compound exhibiting both anti-inflammatory and antifungal properties. Notably flutrimazole demonstrates limited transdermal penetration making it particularly advantageous for topical applications in the treatment of fungal infections especially those accompanied by inflammation. purity: 98%
Catalog No. 50-257-9544