Learn More
Cayman Chemical Fluvastatin Inhibitors
SDP
Supplier:  Cayman Chemical 1001033450 MG
Encompass_Preferred
An HMG-CoA reductase inhibitor (Ki = 0.3 nM for the rat enzyme); inhibits CYP2C9 (IC50 = 100 nM); inhibits oxidized LDL-induced ferroptosis and reverses oxidized LDL-induced decreases in GPX4 and system Xc- levels in HUVECs; decreases serum cholesterol, triglyceride, and phospholipid levels, the formation of TBARS, and vascular ACE activity in rabbits fed a high-cholesterol diet at 2 mg/kg per day; increases survival in a mouse model of myocardial infarction at 10 mg/kg per day
Catalog No. 50-288-8801