Learn More
Apexbio Technology LLC Fluvoxamine 54739-18-3 50mg
SDP
Supplier:  Apexbio Technology LLC B120550
Encompass_Preferred
Fluvoxamine (CAS 54739-18-3) is an antidepressant small molecule that functions primarily by selectively inhibiting serotonin (5-HT) reuptake In vitro studies have shown its ability to inhibit 5-HT uptake in platelets and brain synaptosomes In animal models fluvoxamine administration elevates extracellular serotonin levels across various brain regions and in combination with quetiapine also elevates dopamine levels in specific areas including prefrontal cortex and thalamus Additionally fluvoxamine modulates synaptic plasticity and displays potential analgesic effects mediated via serotonin receptor subtypes (5-HT1A 5-HT2A/2C and 5-HT3) Due to this pharmacological profile fluvoxamine serves as a valuable tool in neurological and psychiatric research
Catalog No. 50-000-62196