Learn More
Medchemexpress LLC Fluvoxamine | 54739-18-3 | 99.4% | C15H21F3N2O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB010350MG
Encompass_Preferred
Fluvoxamine (DU-23000) is an antidepressant that functions pharmacologically as a selective serotonin reuptake inhibitor (SSRI). It is effective in inhibiting 5-ht uptake by blood platelets and brain synaptosomes.
  • Antagonism of the reserpine-induced lowering of the pentamethylenetetrazole convulsive threshold suggests an effect on 5-HT uptake.
  • Does not produce stimulatory effects in rats when rapidly acting reserpine-like compounds are administered after a dose.
  • Has shown potential to improve combat-related PTSD symptoms, although controlled studies with a placebo group are warranted.
  • Potency in reducing ethanol self-administration depends on whether ethanol is available in isolation or concurrently with food reinforcement.
Catalog No. 50-004-32963