Learn More
Apexbio Technology LLC Fluvoxamine maleate 61718-82-9 10mg
SDP
Supplier:  Apexbio Technology LLC A255310
Encompass_Preferred
Fluvoxamine maleate (CAS 61718-82-9) is a selective serotonin reuptake inhibitor that acts by inhibiting the human serotonin (5-HT) transporter This action leads to increased extracellular serotonin concentrations Fluvoxamine maleate exhibits high binding affinity for the 5-HT transporter with a reported K sub i /sub of 1 6 nmol/l It is widely utilized in biomedical research to investigate serotonergic signaling pathways and to model mechanisms underlying antidepressant activity Fluvoxamine maleate is also available as part of the Serotonin Uptake Inhibitor Tocriset for experimental applications
Catalog No. 50-000-94918