Learn More
Medchemexpress LLC Fluvoxamine maleate | 61718-82-9 | 99.8% | C19H25F3N2O6 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYB0103A10MM/1ML
Encompass_Preferred
Fluvoxamine maleate is an antidepressant that functions pharmacologically as a selective serotonin reuptake inhibitor. It is effective in inhibiting 5-HT uptake by blood platelets and brain synaptosomes. It appears to improve combat-related PTSD symptoms, although findings are limited by high attrition rates and the absence of a placebo group in some studies.
  • Functions as a selective serotonin reuptake inhibitor
  • Effective in inhibiting 5-HT uptake by blood platelets and brain synaptosomes
  • Antagonizes reserpine-induced lowering of pentamethylenetetrazole convulsive threshold
  • Does not produce stimulatory effects in rats when rapidly acting reserpine-like compounds are administered after a dose
  • Potency in reducing ethanol-maintained behavior varies depending on whether ethanol is available in isolation or concurrently with food
Catalog No. 50-004-32828