Learn More
Medchemexpress LLC Fluvoxamine maleate | 61718-82-9 | 99.8% | C19H25F3N2O6 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB01035MG
Encompass_Preferred
Fluvoxamine maleate is an antidepressant that operates pharmacologically as a selective serotonin reuptake inhibitor. It is effective in inhibiting 5-HT uptake by blood platelets and brain synaptosomes, and it has been studied for its potential in treating various conditions, including depressive disorder, obsessive-compulsive disorder, and post-traumatic stress disorder.
  • Functions as an antidepressant.
  • Acts as a selective serotonin reuptake inhibitor.
  • Inhibits serotonin uptake by blood platelets and brain synaptosomes.
  • Demonstrates spermicidal activity against human spermatozoa.
  • Shown in studies to improve PTSD symptoms.
  • Has been investigated for treating depressive and obsessive-compulsive disorders.
Catalog No. 50-004-32827