Learn More
Medchemexpress LLC Fluzoparib 10mg | 1358715-18-0 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11477810MG
Encompass_Preferred
Fluzoparib is a potent, orally active PARP-1 inhibitor used for preclinical research into DNA repair and PARP-targeted cancer therapies. It shows sub-nanomolar enzymatic potency and has demonstrated activity in homologous recombination-deficient cancer models, with practical formulations for laboratory use.
  • Potent PARP-1 inhibition (IC50 ≈ 1.46 nM)
  • Selective activity against homologous recombination-deficient cells
  • Sensitizes cells to cytotoxic agents
  • Suitable for in vitro and in vivo preclinical studies
  • Available as small solid quantities and DMSO solution formats
Catalog No. 50-002-27376