Learn More
Thomas Scientific Fly bottle, square bottom polypropylene, 6 oz, 500/case
Supplier: Thomas Scientific 1158J85
Parter Medical Products manufactures Drosophila products that offer flexibility and product excellence at the best price. These products are specifically designed for Drosophila research. Manufactured from pure virgin medical grade plastic Quality Design Dependable Service Bottles are re-usable yet disposably priced Bottles Available in transparent autoclavable polypropylene or less transparent non-autoclavable polyethylene Bottle openings are designed to accept statndard size paper lid, foam plugs, cotton balls and Buzz Plugs™