Learn More
CELLTREAT Scientific Products FlyTreat™ Drosophila Vial, Wide Opening, Polypropylene, Bulk, Non-sterile, Case Quantity 500
Supplier: CELLTREAT Scientific Products 229952
Crystal-clear polystyrene vials provide a controlled environment forpopulations of fruit flies to feed, mate, and lay eggs
Polypropylene vials are durable and autoclavable
Designed with a straight wall and flat bottom for easy filling, plugging,and racking in trays
Safe, lighter, and more economical alternative to traditional glass vials
Available in bulk packs and convenient tray packs