Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Female Luer to Hose Barb Adapter, 1/4"" ID; 25/PK
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX3080010
Encompass
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.
Catalog No. NC0287215