Learn More
Biotium Forget-Me-Not™ Universal Probe Master Mix (100 reactions)
Supplier:  Biotium 31043T
Encompass_Preferred
2X Forget-Me-Not™ Universal qPCR Probe Master Mix is a high-performance product for fluorescent probe-based PCR applications, including quantitation and SNP genotyping. This kit is suitable for all fluorescent probe-based technologies, including hydrolysis probes (such as TaqMan and dual-labeled BHQ probes) and displacement probes (like Molecular Beacons). Forget-Me-Not™ Universal Probe Master Mix shows excellent concordance of results in singleplex and multiplex reactions, has broad instrument compatibility, and can be used in both standard and fast protocols. This kit does not contain ROX reference dye. This trial size has sufficient reagent for 100 PCR reactions.
Catalog No. 50-196-4560