Learn More
Cayman Chemical Fmoc-1-Methyl-L-hIstIdIn 50mg
SDP
Supplier:  Cayman Chemical 3631250 MG
Encompass_Preferred
An amino acid-containing building block; has been used in the production of ε-nitrogen-ligated copper centers in copper nitrite reductases
Catalog No. 50-291-2931