Learn More
Medchemexpress LLC Fmoc-1-methyl-L-histidine | 202920-22-7 | MFCD02259495 | 99.2% | 391.42 g·mol⁻1 | C22H21N3O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW04868210MG
Encompass_Preferred
Fmoc-1-methyl-L-histidine is an Fmoc-protected amino acid building block used in peptide synthesis and as an intermediate for biochemical research. It is suitable for incorporation of methylated histidine residues in peptides and for studies of metal-ligated histidine sites.
  • Fmoc-protected amino acid building block.
  • Suitable for Fmoc solid-phase peptide synthesis.
  • Used as an intermediate in biochemical and metalloprotein studies.
  • High purity: 99.2% (HPLC reported).
  • Available as solid and as 10 mM solution in DMSO.
Catalog No. 50-002-10316