Learn More
Medchemexpress LLC Fmoc-12-aminododecan 5g
SDP
Supplier:  Medchemexpress LLC HYW0971285G
Encompass_Preferred
Fmoc-12-aminododecanoic acid is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups The compound can be used as a PROTAC linker in the synthesis of PROTACs The Fmoc group can be deprotected under basic condition to obtain the free amine which can be used for further conjugations The terminal carboxylic acid can react with primary amine groups in the presence of activators (e g EDC or HATU) to form a stable amide bond
Catalog No. 50-002-04512