Learn More
Medchemexpress LLC Fmoc-3-ala(3-thienyl)-oh | 186320-06-9 | MFCD00151916 | 99.8% | 393.46 g/mol | C22H19NO4S | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0160321G
Encompass_Preferred
Fmoc-3-Ala(3-thienyl)-OH is an Fmoc-protected amino acid derivative (an alanine analogue bearing a 3-thienyl/thiophene side chain) used as a building block in solid-phase peptide synthesis and related peptide chemistry. It is provided at research-grade purity and is employed when introducing a thiophene-containing residue into peptides for structure-activity studies.
  • Fmoc-protected amino acid for Fmoc-based peptide synthesis.
  • 3-thienyl side chain introduces a thiophene moiety into peptides.
  • Useful as a noncanonical residue for structure-activity investigations.
  • High purity (99.8%) suitable for analytical and synthetic applications.
  • Available in multiple pack sizes to support small-scale synthesis.
  • Molecular weight 393.46 g/mol; chemical formula C22H19NO4S.
Catalog No. 50-003-00585