Learn More
STA PHARMACEUTICAL US LLC Fmoc-3-amino-3-(1-Naphthyl)propionic acid | 100 g | CAS 269078-77-5 | MDL MFCD02090642
Supplier: STA PHARMACEUTICAL US LLC STAAA10093100GM
Fmoc-3-amino-3-(1-Naphthyl)propionic acid is a Amino Acid reagent (Subcategory: Beta AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 269078-77-5
- MDL: MFCD02090642
- InChIKey: YQHVLHLQDDNYKX-UHFFFAOYSA-N
- Molecular Weight: 437.495
- Molecular Formula: C28H23NO4
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(naphthalen-1-yl)propanoic acid
- SMILES: O=C(NC(C1=C(C=CC=C2)C2=CC=C1)CC(O)=O)OCC3C(C=CC=C4)=C4C5=C3C=CC=C5
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.