Learn More
STA PHARMACEUTICAL US LLC Fmoc-3-styryl-L-alanine | 100 g | CAS 159610-82-9 | MDL MFCD01311748
Supplier: STA PHARMACEUTICAL US LLC STAAA10672100GM
Fmoc-3-styryl-L-alanine is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 159610-82-9
- MDL: MFCD01311748
- InChIKey: ZFMHHKMOLFNMMV-DDVUFSRBSA-N
- Molecular Weight: 413.473
- Molecular Formula: C26H23NO4
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (S,E)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-5-phenylpent-4-enoic acid
- SMILES: O=C([C@H](C/C=C/C1=CC=CC=C1)NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.