Learn More
Medchemexpress LLC Fmoc-3VVD-OH | 863971-44-2 | 100.7% | 637.81 g/mol | C36H51N3O7 | 1 G

Supplier: Medchemexpress LLC HY789211G
Fmoc-3VVD-OH is an Fmoc-protected, cleavable antibody-drug conjugate (ADC) linker supplied for research use and analytical applications where a protected carboxylic acid linker is required.
- Cleavable linker suitable for ADC synthesis.
- Fmoc-protected carboxylic acid for linker and peptide chemistry.
- CAS number 863971-44-2.
- Molecular weight 637.81 g/mol.
- Reported purity 100.7% (as listed on product page).
- Available in multiple pack sizes, including 1 G.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.