Learn More
Medchemexpress LLC Fmoc-3VVD-OH | 863971-44-2 | MFCD28386982 | >95.0% | 637.81 g/mol | C36H51N3O7 | 10 MG

Supplier: Medchemexpress LLC HY78921R10MG
Fmoc-3VVD-OH is an Fmoc-protected ADC linker analytical standard supplied as a solid reference material for analytical method development and quantitative analysis by LC, GC, and MS. It is intended for research and analytical applications in bioconjugation and ADC synthesis workflows.
- Analytical standard for LC, GC, and MS method development.
- Intended for use in ADC linker synthesis and analytical testing.
- CAS number 863971-44-2 and molecular formula C36H51N3O7.
- Molecular weight approximately 637.81 g/mol.
- Purity greater than 95% for the standard product.
- Provided as a solid in small pack sizes for analytical use (e.g., 10 MG).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.