Learn More
Medchemexpress LLC Fmoc-3VVD-OH (Standard) | 863971-44-2 | 100.7% | C36H51N3O7 | 100 MG

Supplier: Medchemexpress LLC HY78921R100MG
Fmoc-3VVD-OH (Standard) is an analytical standard and a cleavable ADC linker utilized in the synthesis of antibody-drug conjugates (ADCs). With a molecular weight of 637.81 and a chemical formula of C36H51N3O7, it presents as a white to off-white solid. This product is designed for research and analytical applications.
- Functions as a cleavable ADC linker
- Used for synthesizing antibody-drug conjugates (ADCs)
- Serves as an analytical standard
- Intended for research and analytical use
- Store at 4°C, protected from light
- For solutions, store at -80°C for 6 months or -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.