Learn More
STA PHARMACEUTICAL US LLC Fmoc-5-Methoxy-D-Tryptophan | 1 g | CAS 1353018-42-4 | MDL MFCD28166523
Supplier: STA PHARMACEUTICAL US LLC STAAA114491GM
Fmoc-5-Methoxy-D-Tryptophan is a Amino Acid reagent (Subcategory: Trp) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1353018-42-4
- MDL: MFCD28166523
- InChIKey: LOSXXKTXEHICBG-RUZDIDTESA-N
- Molecular Weight: 456.498
- Molecular Formula: C27H24N2O5
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29339980
- Country Of Origin: China
- IUPAC: (R)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(5-methoxy-1H-indol-3-yl)propanoic acid
- SMILES: COC1=CC2=C(NC=C2C[C@H](C(O)=O)NC(OCC3C4=CC=CC=C4C5=C3C=CC=C5)=O)C=C1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.