Learn More
Medchemexpress LLC Fmoc-7-amino-heptanoic acid | 127582-76-7 | 98.75% | 367.44 | 5 G
Supplier: Medchemexpress LLC HYW0090565G
Fmoc-7-amino-heptanoic acid is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups. It can be used as a PROTAC linker in the synthesis of PROTACs.
- Fmoc group can be deprotected under basic conditions to obtain the free amine for further conjugations.
- Terminal carboxylic acid can react with primary amine groups in the presence of activators (e.g., EDC or HATU) to form a stable amide bond.