Learn More
Bachem Fmoc-8-aminooctanoic acid
SDP
Supplier:  Bachem 4045043.0005
Encompass_Preferred
Fmoc-8-aminooctanoic acid, 126631-93-4, C23H27NO4, Mr 381.47, 5g. Fmoc-o-aminocaprylic acid, Fmoc-8-Aoc-OH. Aoc, a flexible hydrophobic spacer, can also be incorporated in a peptide chain as an equivalent of a tripeptide.
Catalog No. 50-258-811