Learn More
eMolecules​ Fmoc-a-Me-D-leu-OH | Chem-Impex | 1231709-23-1 | MFCD02682477 | 367.445 | C22H25NO4 | 98.000 | CC(C)CC(C)(NC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 1g | 272389301
Supplier:  eMolecules​ 272389301
Encompass_Preferred
Fmoc-a-Me-D-leu-OH | Chem-Impex | 1231709-23-1 | MFCD02682477 | 367.445 | C22H25NO4 | 98.000 | CC(C)CC(C)(NC(=O)OCC1c2ccccc2-c2ccccc12)C(O)=O | 1g | 272389301
Catalog No. NC1724857