Learn More
Medchemexpress LLC Fmoc-Abu-OH | 135112-27-5 | 99.2% | 325.36 | 25 G

Supplier: Medchemexpress LLC 25GHYW009151
Fmoc-Abu-OH is an alanine derivative. Amino acids and their derivatives are commercially used as ergogenic supplements, influencing the secretion of anabolic hormones, fuel supply during exercise, mental performance during stress, and preventing exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
- Solid appearance
- White to off-white color
- Stable as powder for up to 3 years at -20°C or 2 years at 4°C
- Stable in solvent for up to 6 months at -80°C or 1 month at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.