Learn More
STA PHARMACEUTICAL US LLC Fmoc-Aeg-OAll HCl | 5 g | CAS 861114-47-8 | MDL MFCD28404628
Supplier: STA PHARMACEUTICAL US LLC STAAA114335GM
Fmoc-Aeg-OAll HCl is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 861114-47-8
- MDL: MFCD28404628
- InChIKey: XFOMYRTWUSNKBU-UHFFFAOYSA-N
- Molecular Weight: 416.902
- Molecular Formula: C22H25ClN2O4
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: allyl (2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)ethyl)glycinate hydrochloride
- SMILES: C=CCOC(CNCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)=O.Cl
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.