Learn More
Bachem Fmoc-Aib-OH
SDP
Supplier:  Bachem 4013932.0025
Encompass_Preferred
Fmoc-Aib-OH, 94744-50-0, C19H19NO4, Mr 325.36, 25g. Fmoc-a-Me-Ala-OH. The acid fluoride of this sterically hindered amino acid could be coupled to N-methylated amino acids in HFIP at 55°C.
Catalog No. 50-258-588