Learn More
Medchemexpress LLC Fmoc-Ala-Ala-PAB | 1949793-44-5 | 97.3% | 458.51 g/mol | C23H30N4O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14846410MG
Encompass_Preferred
MC-Ala-Ala-PAB is a cleavable antibody-drug conjugate (ADC) linker containing an Ala-Ala dipeptide and a para-aminobenzyl (PAB) self-immolative spacer. The N-terminus is Fmoc-protected to facilitate incorporation during solid-phase or solution-phase synthesis of ADCs.
  • Provides protease-cleavable release of payloads via an Ala-Ala dipeptide spacer.
  • Contains a para-aminobenzyl (PAB) self-immolative linker.
  • Fmoc-protected N-terminus for compatibility with peptide synthesis workflows.
  • Supplied with analytical characterization such as HPLC, LC-MS, and HNMR.
  • High purity suitable for research synthesis (purity 97.3%).
Catalog No. 50-002-02653