Learn More
Medchemexpress LLC Fmoc-Ala-Ala-PAB | 1949793-44-5 | 97.3% | 458.51 g/mol | C23H30N4O6 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY14846425MG
Encompass_Preferred
MC-Ala-Ala-PAB is a cleavable protease-sensitive linker (Fmoc-Ala-Ala-PAB) used in antibody-drug conjugate synthesis. The Fmoc-protected dipeptide (Ala-Ala) combined with a para-aminobenzyl (PAB) self-immolative spacer enables intracellular payload release upon enzymatic cleavage. The reagent is supplied at high purity for research and conjugation workflows.
  • Cleavable dipeptide designed for protease-sensitive payload release.
  • Fmoc-protected for compatibility with solid-phase and solution-phase synthesis.
  • Para-aminobenzyl self-immolative spacer for efficient release of linked payloads.
  • High purity suitable for conjugation and analytical workflows.
  • Available in small-milligram pack sizes for research use.
Catalog No. 50-002-02555