Learn More
Medchemexpress LLC Fmoc-Ala-OH-13C3 | 765259-05-0 | 98.5% | 314.31 | 25 MG
Supplier: Medchemexpress LLC 25MGHYW009204S5
Fmoc-Ala-OH-13C3 is a 13C-labeled alanine derivative. This compound serves as both a tracer and an internal standard for various analytical applications.
- Can be used for quantitative analysis by NMR, GC-MS, or LC-MS.
- Contains stable heavy isotopes incorporated into drug molecules.
- Functions as a tracer for quantitation during drug development processes.
- Has the potential to influence pharmacokinetic and metabolic profiles of drugs through deuteration.