Learn More
Medchemexpress LLC Fmoc-Ala-OH-13C3 | 765259-05-0 | 98.5% | 314.31 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW009204S55MG
Encompass_Preferred
Fmoc-Ala-OH-13C3 is a 13C-labeled derivative of Fmoc-Ala-OH, which itself is an alanine derivative. This compound is intended for research use only and is not sold to patients. It can be utilized as a tracer and as an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Functions as an internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, or LC-MS applications.
Catalog No. 50-004-53177