Learn More
Medchemexpress LLC Fmoc-Ala-OH-3-13C | 201489-21-6 | 98.8% | 312.32 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW009204S3100MG
Encompass_Preferred
Fmoc-Ala-OH-3-13C is the 13C labeled Fmoc-Ala-OH. It is for research use only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • 13C labeled compound
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs through deuteration
Catalog No. 50-004-53573