Learn More
Medchemexpress LLC Fmoc-Ala-OH-3-13C | 201489-21-6 | 98.8% | 312.32 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW009204S350MG
Encompass_Preferred
Fmoc-Ala-OH-3-13C is the 13C labeled Fmoc-Ala-OH, intended for research use only. This compound is part of a class of stable heavy isotopes incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration, in particular, has shown potential in affecting pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-53750