Learn More
Apexbio Technology LLC Fmoc-Ala-OMe 146346-88-5 5g
SDP
Supplier:  Apexbio Technology LLC A73015000
Encompass_Preferred
Fmoc-Ala-OMe is an alanine-based amino acid derivative functionalized with an N-terminal Fmoc protecting group and a methyl ester at its carboxyl terminus The introduction of the Fmoc moiety enables selective deprotection under mild basic conditions facilitating stepwise peptide assembly in solid-phase peptide synthesis (SPPS) The methyl ester serves as a stable carboxyl protective group that ensures intermediate peptide derivatives maintain structural integrity during synthetic procedures Fmoc-Ala-OMe is utilized primarily in constructing peptides for structural and functional characterization enabling studies on structure-activity relationships conformational analysis and peptide bioactivity assessment
Catalog No. 50-224-0849