Learn More
Apexbio Technology LLC Fmoc-Arg-OH 91000-69-0 100g
SDP
Supplier:  Apexbio Technology LLC A7306100000
Encompass_Preferred
Fmoc-Arg-OH (91000-69-0) is an Fmoc-protected derivative of the amino acid L-arginine regularly employed in solid-phase peptide synthesis (SPPS) The molecule possesses an Fmoc protecting group on the amino functionality enabling selective and controlled peptide chain elongation Due to its guanidinium side chain arginine residues participate in characteristic ionic interactions and hydrogen bonds within peptides and proteins Based on these properties Fmoc-Arg-OH holds research potential in peptide and protein biochemistry including studies of protein structure and function signaling pathways receptor-ligand interactions pharmacologically relevant peptide development and structure-activity relationship investigations
Catalog No. 50-000-93526