Learn More
Medchemexpress LLC Fmoc-Arg-OH | 91000-69-0 | 99.4% | 396.44 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW01375010MM/1ML
Encompass_Preferred
Fmoc-Arg-OH (Fmoc-L-Arginine) is an Fmoc modified Arginine used in peptide synthesis. It is a key raw material in the solid-phase synthesis of peptides containing arginine residues. Optimized conditions for its attachment to the Wang polymer can reduce side reactions.
  • Used in peptide synthesis
  • Key raw material for solid-phase synthesis of arginine-containing peptides
  • Optimized conditions reduce side reactions
  • Plays an important role in the synthesis of bradykinin
  • Plays an important role in the synthesis of octapeptides related to the transmembrane gp41 of HIV-1
Catalog No. 50-004-54199