Learn More
Medchemexpress LLC Fmoc-Arg-OH (Fmoc-L-Arginine) | 91000-69-0 | 99.35% | 396.44 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW013750100G
Encompass_Preferred
Fmoc-Arg-OH (Fmoc-L-Arginine) is an Fmoc modified Arginine used in peptide synthesis. It is a key raw material in the solid-phase synthesis of peptides containing arginine residues. The optimized conditions for its attachment to the Wang polymer can reduce side reactions. Fmoc-Arg-OH plays an important role in the synthesis of bradykinin and octapeptides related to the transmembrane gp41 of HIV-1.
  • Fmoc modified arginine
  • Used in peptide synthesis
  • Key raw material in solid-phase synthesis of peptides containing arginine residues
  • Optimized conditions for attachment to Wang polymer reduce side reactions
  • Important for synthesis of bradykinin and octapeptides related to transmembrane gp41 of HIV-1
Catalog No. 50-004-54703