Learn More
Medchemexpress LLC Fmoc-Arg-OH (Fmoc-L-Arginine) | 91000-69-0 | 99.95% | 396.44 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01375010G
Encompass_Preferred
Fmoc-Arg-OH (Fmoc-L-Arginine) is an Fmoc modified Arginine used in peptide synthesis. It is a key raw material for the solid-phase synthesis of peptides containing arginine residues. Optimized conditions for its attachment to the Wang polymer can help reduce side reactions. This compound is also important in the synthesis of bradykinin and octapeptides related to the transmembrane gp41 of HIV-1.
  • Fmoc modified Arginine
  • Used in peptide synthesis
  • Key raw material for solid-phase synthesis of peptides containing arginine residues
  • Plays an important role in the synthesis of bradykinin and octapeptides related to the transmembrane gp41 of HIV-1
Catalog No. 50-004-54494