Learn More
Cayman Chemical Fmoc-Arg Pbf-OH 1g
SDP
Supplier:  Cayman Chemical 364581 G
Encompass_Preferred
An amino acid-containing building block; has been used in solid-phase peptide synthesis
Catalog No. 50-291-2487