Learn More
Bachem Fmoc-Arg(Pbf)-OH
SDP
Supplier:  Bachem 4089151.0025
Encompass_Preferred
Fmoc-Arg(Pbf)-OH, 154445-77-9, C34H40N4O7S, Mr 648.78, 25g. Pbf protection is especially recommended for multiple-Arg- and Trp-containinng peptides. TFA cleavage is best performed in the presence of a silane scavenger.
Catalog No. 50-258-639