Learn More
Bachem Fmoc-Arg(Tos)-OH
SDP
Supplier:  Bachem 4004138.0025
Encompass_Preferred
Fmoc-Arg(Tos)-OH, 83792-47-6, C28H30N4O6S, Mr 550.64, 25g. Fmoc-Arg(Tos) was shown to inhibit HIV-1 protease (IC50 5 μM) and cathepsin D (IC50 12 μM).
Catalog No. 50-194-6953