Learn More
Medchemexpress LLC Fmoc-asn-pro-val-pabc-pnp | 2893871-64-0 | 99.3% | 820.84 g·mol⁻1 | C43H44N6O11 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1446285MG
Encompass_Preferred
Fmoc-Asn-Pro-Val-PABC-PNP is an Fmoc-protected peptide-based ADC linker for research applications. It incorporates a para-aminobenzyl carbamate (PABC) spacer and a p-nitrophenyl (PNP) activated leaving group to facilitate conjugation and controlled payload release during antibody-drug conjugate synthesis.
  • Peptide-based linker suitable for ADC synthesis.
  • Contains PABC spacer for efficient payload release.
  • PNP-activated leaving group enables facile conjugation chemistry.
  • Supplied as a solid with high analytical purity for research use.
  • Available in small milligram quantities for laboratory experiments.
Catalog No. 50-002-21560